5-(Trifluoromethyl)-2,3-dihydro-1H-inden-1-one 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Trifluoromethyl)indan-1-one

PC8805-1G PC8805-250MG PC8805-5G
APOSPC8805-1GEA 192 EUR
APOSPC8805-1G APOSPC8805-250MG APOSPC8805-5G
5-(Trifluoromethyl)-2,3-dihydro-1H-inden-1-one 98%
5-(Trifluoromethyl)-2,3-dihydro-1H-inden-1-one
Formel: C₁₀H₇F₃O
Molecular Weight: 200.1572 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07772122
CAS-Nummer: 150969-56-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR