Ethyl-2-fluoro-5-nitrobenzenecarboxylate 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-fluoro-5-nitrobenzoate

PC8810-500MG
APOSPC8810-500MGEA 0 EUR
APOSPC8810-500MG
Ethyl-2-fluoro-5-nitrobenzenecarboxylate 95+%
Ethyl-2-fluoro-5-nitrobenzenecarboxylate
Formel: C₉H₈FNO₄
Molecular Weight: 213,17 g/mol
Schmelzpunkt: 53…55 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06204381
CAS-Nummer: 367-79-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR