6-(Trifluoromethyl)pyridine-3-amidine hydrochloride 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-(Trifluoromethyl)nicotinimidamide hydrochloride, 6-(Trifluoromethyl)nicotinamidine hydrochloride
PC8819-1G
PC8819-5G
APOSPC8819-1GEA
60.9
EUR
APOSPC8819-1G
APOSPC8819-5G
6-(Trifluoromethyl)pyridine-3-amidine hydrochloride 99%
6-(Trifluoromethyl)pyridine-3-amidine hydrochloride
Formel:
C₇H₇ClF₃N₃ Molecular Weight: 225,6 g/mol Schmelzpunkt: 191…193 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01862652 CAS-Nummer: 221313-11-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...