2-Brom-3-fluorpyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC8846-5G
PC8846-100G
PC8846-25G
APOSPC8846-5GEA
21.9
EUR
APOSPC8846-5G
APOSPC8846-100G
APOSPC8846-25G
2-Brom-3-fluorpyridin 98%
2-Bromo-3-fluoropyridine
Formel:
C₅H₃BrFN Dichte: 1,74 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD04114128 CAS-Nummer: 40273-45-8 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...