4-(4-Fluorophenyl)-6-hydroxy-2-phenyl-5-pyrimidinecarbonitrile
Lieferant: Apollo Scientific
PC8921-10G
PC8921-1G
PC8921-5G
APOSPC8921-10GEA
546
EUR
APOSPC8921-10G
APOSPC8921-1G
APOSPC8921-5G
4-(4-Fluorophenyl)-6-hydroxy-2-phenyl-5-pyrimidinecarbonitrile
4-(4-Fluorophenyl)-6-hydroxy-2-phenyl-5-pyrimidinecarbonitrile
Formel:
C₁₇H₁₀FN₃O Molecular Weight: 291.29 g/mol Schmelzpunkt: 330 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00794103 CAS-Nummer: 477853-95-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...