(4-Brom-2-fluorphenyl)acetonitril
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-2-fluorobenzyl cyanide, 2-(4-Bromo-2-fluorophenyl)acetonitrile
PC8933-100G
PC8933-25G
PC8933-5G
APOSPC8933-100GEA
127
EUR
APOSPC8933-100G
APOSPC8933-25G
APOSPC8933-5G
(4-Brom-2-fluorphenyl)acetonitril
(4-Brom-2-fluorphenyl)acetonitril
Formel:
C₈H₅BrFN Molecular Weight: 214,0300 g/mol Schmelzpunkt: 52…53 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368653 CAS-Nummer: 114897-91-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...