(3R,4R)-1-Benzyl-4-(trifluoromethyl)tetrahydro-1H-pyrrole-3-carboxylic acid 95+%

Lieferant: Apollo Scientific

Synonyme: (3R,4R)-1-Benzyl-4-(trifluoromethyl)pyrrolidine-3-carboxylic acid

PC8940-1G PC8940-500MG
APOSPC8940-1GEA 502 EUR
APOSPC8940-1G APOSPC8940-500MG
(3R,4R)-1-Benzyl-4-(trifluoromethyl)tetrahydro-1H-pyrrole-3-carboxylic acid 95+%
(3R,4R)-1-Benzyl-4-(trifluoromethyl)tetrahydro-1H-pyrrole-3-carboxylic acid
Formel: C₁₃H₁₄F₃NO₂
Molecular Weight: 273.26 g/mol
Schmelzpunkt: 92 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD12974801
CAS-Nummer: 184844-96-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR