3-Fluor-5-hydroxypyridin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluor-5-pyridinol

PC8941-1G PC8941-5G PC8941-250MG
APOSPC8941-1GEA 39.3 EUR
APOSPC8941-1G APOSPC8941-5G APOSPC8941-250MG
3-Fluor-5-hydroxypyridin 97%
3-Fluor-5-hydroxypyridin
Formel: C₅H₄FNO
Molecular Weight: 113,09 g/mol
Schmelzpunkt: 161…164 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03095199
CAS-Nummer: 209328-55-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR