3-Fluorazetidinhydrochlorid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Fluoroazetidine HCl
PC8943-1G
PC8943-500MG
APOSPC8943-1GEA
24.8
EUR
APOSPC8943-1G
APOSPC8943-500MG
3-Fluorazetidinhydrochlorid 97%
3-Fluoroazetidine hydrochloride
Formel:
C3H7ClFN Molecular Weight: 111,55 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663715 CAS-Nummer: 617718-46-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...