3,5-Difluor-4-hydroxybenzaldehyd 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC8951-1G
PC8951-5G
PC8951-25G
APOSPC8951-1GEA
43.5
EUR
APOSPC8951-1G
APOSPC8951-5G
APOSPC8951-25G
3,5-Difluor-4-hydroxybenzaldehyd 99%
3,5-Difluoro-4-hydroxybenzaldehyde
Formel:
C₇H₄F₂O₂ Schmelzpunkt: 119…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797917 CAS-Nummer: 118276-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...