6-Chloro-3-(trifluoromethyl)pyridine-2-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

PC8967-50MG PC8967-500MG
APOSPC8967-50MGEA 94.5 EUR
APOSPC8967-50MG APOSPC8967-500MG
6-Chloro-3-(trifluoromethyl)pyridine-2-carboxylic acid 97%
6-Chloro-3-(trifluoromethyl)pyridine-2-carboxylic acid
Formel: C₇H₃ClF₃NO₂
Molecular Weight: 225,5500 g/mol
Schmelzpunkt: 109…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08235206
CAS-Nummer: 796090-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR