2-(Trifluormethyl)isonicotinsäure 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Trifluoromethyl)pyridine-4-carboxylic acid
PC8970-1G
PC8970-5G
APOSPC8970-1GEA
58
EUR
APOSPC8970-1G
APOSPC8970-5G
2-(Trifluormethyl)isonicotinsäure 96%
2-(Trifluoromethyl)isonicotinic acid
Formel:
C₇H₄F₃NO₂ Molecular Weight: 191,11 g/mol Schmelzpunkt: 217…223 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07774132 CAS-Nummer: 131747-41-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...