2-Hydrazino-4-(trifluoromethyl)pyridine 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Hydrazinyl-4-(trifluoromethyl)pyridine
PC8988-100MG
PC8988-1G
PC8988-250MG
APOSPC8988-100MGEA
37.7
EUR
APOSPC8988-100MG
APOSPC8988-1G
APOSPC8988-250MG
2-Hydrazino-4-(trifluoromethyl)pyridine 95%
2-Hydrazino-4-(trifluoromethyl)pyridine
Formel:
C₆H₆F₃N₃ Molecular Weight: 177.13 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08056340 CAS-Nummer: 89570-84-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...