2-Fluoro-N,N-dimethyl-4-nitroaniline 98%

Lieferant: Apollo Scientific

PC900136-10G PC900136-1G PC900136-25G
APOSPC900136-10GEA 364 EUR
APOSPC900136-10G APOSPC900136-1G APOSPC900136-25G
2-Fluoro-N,N-dimethyl-4-nitroaniline 98%
2-Fluoro-N,N-dimethyl-4-nitroaniline
Formel: C₈H₉FN₂O₂
Molecular Weight: 184.17 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 65739-04-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR