4-Fluor-3-methoxybenzylamin 95%
Lieferant: Apollo Scientific
Synonyme:
5-Aminomethyl-2-fluoroanisole, 5-(Aminomethyl)-2-fluoroanisole
PC900427-10G
APOSPC900427-10GEA
991
EUR
APOSPC900427-10G
4-Fluor-3-methoxybenzylamin 95%
4-Fluor-3-methoxybenzylamin
Formel:
C₈H₁₀FNO Molecular Weight: 155.1700 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04116362 CAS-Nummer: 508177-67-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...