2-Chloro-5-fluoroisonicotinaldehyde 98%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-5-fluoropyridine-4-carbaldehyde

PC900544-1G PC900544-250MG
APOSPC900544-1GEA 188 EUR
APOSPC900544-1G APOSPC900544-250MG
2-Chloro-5-fluoroisonicotinaldehyde 98%
2-Chloro-5-fluoroisonicotinaldehyde
Formel: C₆H₃ClFNO
Molecular Weight: 159,55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04972411
CAS-Nummer: 884494-54-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR