3-Brom-5-fluorpyridin-N-oxid 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC900555-1G
PC900555-5G
APOSPC900555-1GEA
190
EUR
APOSPC900555-1G
APOSPC900555-5G
3-Brom-5-fluorpyridin-N-oxid 98%
3-Brom-5-fluorpyridin-N-oxid
Formel:
C₅H₃BrFNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD16036155 CAS-Nummer: 1221793-60-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...