3,5-Difluorpyridin-N-oxid 98%

Lieferant: Apollo Scientific

PC900556-1G PC900556-5G
APOSPC900556-1GEA 168 EUR
APOSPC900556-1G APOSPC900556-5G
3,5-Difluorpyridin-N-oxid 98%
3,5-Difluorpyridin-N-oxid
Formel: C₅H₃F₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD16036156
CAS-Nummer: 210169-07-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR