6-Fluor-2-methoxychinolin 98%
Lieferant: Apollo Scientific
PC900559-1G
PC900559-5G
APOSPC900559-1GEA
145
EUR
APOSPC900559-1G
APOSPC900559-5G
6-Fluor-2-methoxychinolin 98%
6-Fluoro-2-methoxyquinoline
Formel:
C₁₀H₈FNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD16295129 CAS-Nummer: 1226808-76-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...