6-Methyl-2-(trifluoromethyl)pyrimidin-4(3H)-one 95%

Lieferant: Apollo Scientific

Synonyme: 6-Methyl-2-trifluoromethylpyrimidin-4-ol

PC900665-1G PC900665-5G
APOSPC900665-1GEA 270 EUR
APOSPC900665-1G APOSPC900665-5G
6-Methyl-2-(trifluoromethyl)pyrimidin-4(3H)-one 95%
6-Methyl-2-(trifluoromethyl)pyrimidin-4(3H)-one
Formel: C₆H₅F₃N₂O
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2557-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR