4-Chlor-3,5-difluorbenzonitril 95%

Lieferant: Apollo Scientific

PC900666-10G PC900666-25G PC900666-5G
APOSPC900666-10GEA 505 EUR
APOSPC900666-10G APOSPC900666-25G APOSPC900666-5G
4-Chlor-3,5-difluorbenzonitril 95%
4-Chlor-3,5-difluorbenzonitril
Formel: C₇H₂ClF₂N
Molecular Weight: 173,5500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18415564
CAS-Nummer: 144797-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR