2-(2-Fluorophenyl)pyrrolidine 95%

Lieferant: Apollo Scientific

PC900745-1G PC900745-5G PC900745-10G
APOSPC900745-1GEA 127 EUR
APOSPC900745-1G APOSPC900745-5G APOSPC900745-10G
2-(2-Fluorophenyl)pyrrolidine 95%
2-(2-Fluorophenyl)pyrrolidine
Formel: C₁₀H₁₂FN
Lagertemperatur: Raumtemperatur
MDL: MFCD02663474
CAS-Nummer: 72216-04-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR