1-(2,2,2-Trifluoroethyl)-1H-pyrazole-3-carboxylic acid 95%

Lieferant: Apollo Scientific

PC900791-100MG PC900791-1G PC900791-250MG
APOSPC900791-100MGEA 213 EUR
APOSPC900791-100MG APOSPC900791-1G APOSPC900791-250MG
1-(2,2,2-Trifluoroethyl)-1H-pyrazole-3-carboxylic acid 95%
1-(2,2,2-Trifluoroethyl)-1H-pyrazole-3-carboxylic acid
Formel: C₆H₅F₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08556202
CAS-Nummer: 942853-22-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR