Methyl-3-fluor-4-methylbenzoat 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 3-fluoro-4-methylbenzoate
PC900830-10G
PC900830-25G
PC900830-5G
APOSPC900830-10GEA
94.4
EUR
APOSPC900830-10G
APOSPC900830-25G
APOSPC900830-5G
Methyl-3-fluor-4-methylbenzoat 95%
Methyl-3-fluor-4-methylbenzoat
Formel:
C₉H₉FO₂ Molecular Weight: 168,1650 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06203786 CAS-Nummer: 87808-48-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...