7-Fluoro-1H-indole-2-carboxylic acid 98%

Lieferant: Apollo Scientific

PC901298-5G
APOSPC901298-5GEA 94.4 EUR
APOSPC901298-5G
7-Fluoro-1H-indole-2-carboxylic acid 98%
7-Fluoro-1H-indole-2-carboxylic acid
Formel: C₉H₆FNO₂
Molecular Weight: 179,1500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02664429
CAS-Nummer: 399-67-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR