Methyl-3-fluor-5-hydroxybenzoat 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-fluoro-5-hydroxybenzoate

PC901448-1G
APOSPC901448-1GEA 48 EUR
APOSPC901448-1G
Methyl-3-fluor-5-hydroxybenzoat 98%
Methyl-3-fluor-5-hydroxybenzoat
Formel: C₈H₇FO₃
Molecular Weight: 170,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14698043
CAS-Nummer: 1072004-32-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR