4-Fluor-N-methylbenzamid 98%

Lieferant: Apollo Scientific

PC901457-25G
APOSPC901457-25GEA 197 EUR
APOSPC901457-25G
4-Fluor-N-methylbenzamid 98%
4-Fluor-N-methylbenzamid
Formel: C₈H₈FNO
Molecular Weight: 153,1536 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00032531
CAS-Nummer: 701-49-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR