2-Fluor-3-hydroxybenzonitril 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluor-3-hydroxybenzoesäurenitril

PC901463-1G PC901463-250MG
APOSPC901463-1GEA 190 EUR
APOSPC901463-1G APOSPC901463-250MG
2-Fluor-3-hydroxybenzonitril 98%
2-Fluor-3-hydroxybenzonitril
Formel: C₇H₄FNO
Molecular Weight: 137,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09839222
CAS-Nummer: 1000339-24-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR