3-Fluor-N-methylbenzamid 98%
Lieferant: Apollo Scientific
Synonyme:
N-Methyl 3-fluorobenzamide
PC901483-1G
PC901483-25G
PC901483-5G
APOSPC901483-1GEA
145
EUR
APOSPC901483-1G
APOSPC901483-25G
APOSPC901483-5G
3-Fluor-N-methylbenzamid 98%
3-Fluor-N-methylbenzamid
Formel:
C₈H₈FNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD06250758 CAS-Nummer: 701-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...