4-Fluor-N-propylbenzamid 98%

Lieferant: Apollo Scientific

Synonyme: N-Propyl 4-fluorobenzamide

PC901488-10G PC901488-25G PC901488-5G
APOSPC901488-10GEA 270 EUR
APOSPC901488-10G APOSPC901488-25G APOSPC901488-5G
4-Fluor-N-propylbenzamid 98%
4-Fluor-N-propylbenzamid
Formel: C₁₀H₁₂FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD01214137
CAS-Nummer: 349129-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR