2-Ethoxy-4-fluoranilin Hydrochlorid 98%

Lieferant: Apollo Scientific

PC901682-1G PC901682-5G
APOSPC901682-1GEA 183 EUR
APOSPC901682-1G APOSPC901682-5G
2-Ethoxy-4-fluoranilin Hydrochlorid 98%
2-Ethoxy-4-fluoranilin Hydrochlorid
Formel: C₈H₁₁ClFNO
Lagertemperatur: Raumtemperatur
MDL: MFCD04115636
CAS-Nummer: 850568-36-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR