5-Amino-8-fluorchinolin 98%
Lieferant: Apollo Scientific
Synonyme:
8-Fluoroquinolin-5-amine
PC901844-1G
PC901844-250MG
APOSPC901844-1GEA
249
EUR
APOSPC901844-1G
APOSPC901844-250MG
5-Amino-8-fluorchinolin 98%
5-Amino-8-fluoroquinoline
Formel:
C₉H₇FN₂ Molecular Weight: 162,1637 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09042730 CAS-Nummer: 175229-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...