1-(4-Fluorophenyl)cyclobutanecarboxylic acid 97%

Lieferant: Apollo Scientific

PC901939-1G PC901939-5G
APOSPC901939-1GEA 139 EUR
APOSPC901939-1G APOSPC901939-5G
1-(4-Fluorophenyl)cyclobutanecarboxylic acid 97%
1-(4-Fluorophenyl)cyclobutanecarboxylic acid
Formel: C₁₁H₁₁FO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08442159
CAS-Nummer: 151157-46-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR