2-(2-Fluorphenyl)oxazol 98%

Lieferant: Apollo Scientific

PC901975-1G PC901975-25G PC901975-5G
APOSPC901975-1GEA 145 EUR
APOSPC901975-1G APOSPC901975-25G APOSPC901975-5G
2-(2-Fluorphenyl)oxazol 98%
2-(2-Fluorophenyl)oxazole
Formel: C₉H₆FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD15143568
CAS-Nummer: 178672-06-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR