1-(4-Fluorophenyl)cyclobutanecarbonitrile 95%
Lieferant: Apollo Scientific
PC901999-1G
PC901999-5G
APOSPC901999-1GEA
232
EUR
APOSPC901999-1G
APOSPC901999-5G
1-(4-Fluorophenyl)cyclobutanecarbonitrile 95%
1-(4-Fluorophenyl)cyclobutanecarbonitrile
Formel:
C₁₁H₁₀FN Molecular Weight: 175.21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10694662 CAS-Nummer: 405090-30-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...