2-(3-Fluorophenyl)-2-methylpropanoic acid 98%

Lieferant: Apollo Scientific

PC902034-1G PC902034-250MG
APOSPC902034-1GEA 609 EUR
APOSPC902034-1G APOSPC902034-250MG
2-(3-Fluorophenyl)-2-methylpropanoic acid 98%
2-(3-Fluorophenyl)-2-methylpropanoic acid
Formel: C₁₀H₁₁FO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11036911
CAS-Nummer: 93748-20-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR