6-Fluor-3-isochinolinol 98%
Lieferant: Apollo Scientific
PC902157-1G
PC902157-5G
APOSPC902157-1GEA
95.7
EUR
APOSPC902157-1G
APOSPC902157-5G
6-Fluor-3-isochinolinol 98%
6-Fluor-3-isochinolinol
Formel:
C₉H₆FNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD11053873 CAS-Nummer: 51463-15-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...