3-(4-Fluorphenyl)-1,2,4-oxadiazol 98%

Lieferant: Apollo Scientific

PC902202-1G PC902202-25G PC902202-5G
APOSPC902202-1GEA 145 EUR
APOSPC902202-1G APOSPC902202-25G APOSPC902202-5G
3-(4-Fluorphenyl)-1,2,4-oxadiazol 98%
3-(4-Fluorophenyl)-1,2,4-oxadiazole
Formel: C₈H₅FN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD16618955
CAS-Nummer: 887763-94-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR