1-Ethyl-6-fluorbenzimidazol 98%

Lieferant: Apollo Scientific

PC902248-1G PC902248-5G PC902248-25G
APOSPC902248-1GEA 132 EUR
APOSPC902248-1G APOSPC902248-5G APOSPC902248-25G
1-Ethyl-6-fluorbenzimidazol 98%
1-Ethyl-6-fluorobenzimidazole
Formel: C₉H₉FN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12546504
CAS-Nummer: 1187385-87-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR