4-(4-Fluorphenyl)benzonitril 98%
Lieferant: Apollo Scientific
Synonyme:
4'-Fluoro-[1,1'-biphenyl]-4-carbonitrile
PC902385-1G
PC902385-5G
APOSPC902385-1GEA
122
EUR
APOSPC902385-1G
APOSPC902385-5G
4-(4-Fluorphenyl)benzonitril 98%
4-(4-Fluorphenyl)benzonitril
Formel:
C₁₃H₈FN Lagertemperatur: Raumtemperatur |
MDL:
MFCD05981088 CAS-Nummer: 10540-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...