6-Fluor-2-tetralon 95%

Lieferant: Apollo Scientific

Synonyme: 6-Fluoro-3,4-dihydronaphthalen-2(1H)-one

PC902713-1G PC902713-5G
APOSPC902713-1GEA 104 EUR
APOSPC902713-1G APOSPC902713-5G
6-Fluor-2-tetralon 95%
6-Fluor-2-tetralon
Formel: C₁₀H₉FO
Molecular Weight: 164,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02684435
CAS-Nummer: 29419-14-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR