3-Fluor-2-methoxy-6-methylpyridin 95%
Lieferant: Apollo Scientific
Synonyme:
3-Fluoro-2-methoxy-6-methylpyridine, 3-Fluoro-2-methoxy-6-picoline
PC902717-5G
APOSPC902717-5GEA
203
EUR
APOSPC902717-5G
3-Fluor-2-methoxy-6-methylpyridin 95%
3-Fluoro-2-methox-6-methylypyridine
Formel:
C₇H₈FNO Molecular Weight: 141,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095083 CAS-Nummer: 375368-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...