2-Fluor-3-hydroxy-6-picolin 98%

Lieferant: Apollo Scientific

Synonyme: 2-Fluoro-6-methylpyridin-3-ol, 2-Fluoro-3-hydroxy-6-methylpyridine

PC902720-1G PC902720-250MG
APOSPC902720-1GEA 504 EUR
APOSPC902720-1G APOSPC902720-250MG
2-Fluor-3-hydroxy-6-picolin 98%
2-Fluor-3-hydroxy-6-picolin
Formel: C₆H₆FNO
Molecular Weight: 127,1200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095222
CAS-Nummer: 209328-87-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR