2-(4-Fluorophenyl)piperazine 95%
Lieferant: Apollo Scientific
PC902756-1G
PC902756-250MG
APOSPC902756-1GEA
290
EUR
APOSPC902756-1G
APOSPC902756-250MG
2-(4-Fluorophenyl)piperazine 95%
2-(4-Fluorophenyl)piperazine
Formel:
C₁₀H₁₃FN₂ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
65709-33-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...