(R)-β-(4-Fluorphenyl)alaninol 97%

Lieferant: Apollo Scientific

Synonyme: (R)-3-Amino-3-(4-fluorophenyl)propan-1-ol

PC902857-1G PC902857-250MG PC902857-500MG
APOSPC902857-1GEA 596 EUR
APOSPC902857-1G APOSPC902857-250MG APOSPC902857-500MG
(R)-β-(4-Fluorphenyl)alaninol 97%
(R)-β-(4-Fluorphenyl)alaninol
Formel: C₉H₁₂FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD07784088
CAS-Nummer: 228422-47-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR