1-(Methoxymethyl)-2-(trifluormethyl)benzol 96%

Lieferant: Apollo Scientific

PC902958-1G PC902958-5G
APOSPC902958-1GEA 145 EUR
APOSPC902958-1G APOSPC902958-5G
1-(Methoxymethyl)-2-(trifluormethyl)benzol 96%
1-(Methoxymethyl)-2-(trifluoromethyl)benzene
Formel: C₉H₉F₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD11504957
CAS-Nummer: 227948-39-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR