1-(Methoxymethyl)-3-(trifluormethyl)benzol 97%

Lieferant: Apollo Scientific

PC902959-1G PC902959-5G
APOSPC902959-1GEA 168 EUR
APOSPC902959-1G APOSPC902959-5G
1-(Methoxymethyl)-3-(trifluormethyl)benzol 97%
1-(Methoxymethyl)-3-(trifluoromethyl)benzene
Formel: C₉H₉F₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD11504958
CAS-Nummer: 380633-51-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR