1-Fluor-3-iod-5-nitrobenzol 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC903680-1G
PC903680-25G
PC903680-5G
APOSPC903680-1GEA
29
EUR
APOSPC903680-1G
APOSPC903680-25G
APOSPC903680-5G
1-Fluor-3-iod-5-nitrobenzol 99%
1-Fluoro-3-iodo-5-nitrobenzene
Formel:
C₆H₃FINO₂ Molecular Weight: 267 g/mol Schmelzpunkt: 77…79 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007219 CAS-Nummer: 3819-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...