4-Bromo-3,5-difluoropyridine 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC904075-1G
PC904075-250MG
PC904075-5G
APOSPC904075-1GEA
202
EUR
APOSPC904075-1G
APOSPC904075-250MG
APOSPC904075-5G
4-Bromo-3,5-difluoropyridine 98%
4-Bromo-3,5-difluoropyridine
Formel:
C₅H₂BrF₂N Molecular Weight: 193.98 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11505879 CAS-Nummer: 1092352-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...