2-Bromo-3-fluoroisonicotinic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-3-fluoropyridine-4-carboxylic acid

PC904529-1G PC904529-5G
APOSPC904529-1GEA 33.5 EUR
APOSPC904529-1G APOSPC904529-5G
2-Bromo-3-fluoroisonicotinic acid 97%
2-Bromo-3-fluoroisonicotinic acid
Formel: C₆H₃BrFNO₂
Molecular Weight: 219.997 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13185796
CAS-Nummer: 1211530-89-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR